Learn More
Eisco™ Green Slate Metamorphic Rock Specimen
SCI_ED
Catalog No. S27593 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27593 1/Ea.
S27615 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27593 Supplier Eisco™ Supplier No. ESNG0086
Add to Cart
Add to Cart
Green slate is a very lightly metamorphosed and well-foliated specimen. It can be distinguished from it's parent rock (Shale) by the difference in pitch when struck.
  • Correlates well to Next Generation Science Standards Evidence
  • Perfect for lessons on metamorphic rocks and alteration
  • Ideal for identification & classification exercises
Specifications
Product Type Metamorphic Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.9 kg