Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Green Tea Extract, 4:1, Powder, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Description
Spectrum™ Chemical Green Tea Extract, 4:1, Powder
Specifications
Specifications
| CAS Max % | 100% |
| Loss on Drying | 9% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 kg |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction