Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner PROMO
Catalog No. p-7114698
Click to view available options
Neck Finish:
15-425
20-400
22-400
24-400
Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
  • The color green easily identifies PTFE liner system
  • F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
  • Available in smooth and ribbed style
Specifications
Color Green
Material Thermoset
No. per Case 500/Cs.
Packaging Bagged
Liner Material F217, PTFE
Neck Finish 15-425
Product Type Ribbed Thermoset Cap
Closure Material Thermoset
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.