Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes

Collect, transport and process capillary blood. Greiner BioOne™ MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.

Manufacturer: Greiner Bio-One 450480

 View more versions of this product

Catalog No. 22-040-200

  Check Availability
Add to Cart

    Specifications

    Product Type Bio-One VACUETTE MiniCollect Capillary Blood Collection Tube
    Capacity (Metric) 0.5mL
    Additive K2 EDTA
    Color Lavender
    Applications Small volume blood collection
    Closure Included Yes

     

    • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
    • Fit into a 13 x 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
    • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
    • All tubes require MiniCollect funnel or capillary collection device (sold separately)

     

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit