Learn More
Greiner Bio-One Snap Caps
Specifications
Specifications
| Product Type | Snap Cap |
| Material | PET |
| Color | Pink |
| Size | Universal |
| For Use With (Equipment) | Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.