Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GREM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570341 Supplier Invitrogen™ Supplier No. PA570341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Goat: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 93%.

In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GREM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Grem1
Gene Accession No. O35793, O70326
Gene Alias C15DUPq; Cell proliferation-inducing gene 2 protein; CKTSF1B1; colorectal adenoma and carcinoma 1; CRAC1; CRCS4; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; DAND2; Down-regulated in Mos-transformed cells protein; down-regulated in v-mos-transformed cells; Drm; DUP15q; Grem; GREM1; gremlin; gremlin 1; gremlin 1 homolog, cysteine knot superfamily; gremlin 1, cysteine knot superfamily, homolog; gremlin 1, DAN family BMP antagonist; gremlin 1-like protein; gremlin-1; HMPS; HMPS1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2; ld; MPSH; PIG2
Gene Symbols Grem1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat GREM1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23892, 50566
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.