Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GREM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588162
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588162 Supplier Invitrogen™ Supplier No. PA588162
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERKYLKRDWC KTQPLKQTIH EEGCNSRTII NRFCYGQCNS FYIPRHIRKE EGSFQSCSFC KPKKFTTMMV TLNCPELQPP TKKKRVTRVK QCRCISIDLD; Positive Samples: SH-SY5Y, HT-29, A-431, LO2.

In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GREM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Grem1
Gene Accession No. O60565
Gene Alias C15DUPq; Cell proliferation-inducing gene 2 protein; CKTSF1B1; colorectal adenoma and carcinoma 1; CRAC1; CRCS4; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; DAND2; Down-regulated in Mos-transformed cells protein; down-regulated in v-mos-transformed cells; Drm; DUP15q; Grem; GREM1; gremlin; gremlin 1; gremlin 1 homolog, cysteine knot superfamily; gremlin 1, cysteine knot superfamily, homolog; gremlin 1, DAN family BMP antagonist; gremlin 1-like protein; gremlin-1; HMPS; HMPS1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2; ld; MPSH; PIG2
Gene Symbols Grem1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 44-143 of human GREM1 (NP_0011782521).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26585
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.