Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GREM2 (Human) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
Description
GREM2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human GREM2 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).
Specifications
Specifications
| Description | gremlin 2, cysteine knot superfamily, homolog (Xenopus laevis) |
| Cross Reactivity | Human |
| Target Species | Human |
| For Use With (Application) | Quantitative |
| Gene ID (Entrez) | 64388 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store at 4°C for six months. For long term storage store at -20°C. Avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction