Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pseudomonas aeruginosa (Gremubamab Biosimilar) Recombinant Humanized mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542275
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542275 Supplier Invitrogen™ Supplier No. MA542275
Add to Cart
Add to Cart
This item is not returnable. View return policy

Humanized mouse Recombinant Monoclonal Antibody

Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. Isotype of this product is: VH-CH1-VH-V, kappa-CH2-CH3.

Pseudomonas Aeruginosa is a gram-negative, opportunistic pathogen commonly found in soil and water. In humans, P. aeruginosa can infect the urinary tract, respiratory and gastrointestinal system, soft tissues, bones and joints leading to severe systemic infections of immunosuppressed hospitalized patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pseudomonas aeruginosa (Gremubamab Biosimilar)
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias p aeruginosa; P. aeruginosa
Host Species Humanized Mouse
Immunogen Bradyrhizobium (sp.ANU 289) PcrV.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype VH-VH
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.