Learn More
Eisco™ Greywacke Sedimentary Rock Specimen
SCI_ED
Catalog No. S27587 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27587 1/Ea.
S27609 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27587 Supplier Eisco™ Supplier No. ESNG0080
Only null left
Add to Cart
Add to Cart
Graywacke is a sedimentary rock composed of large, angular clasts within a fine-grained clay matrix. Formed during underwater avalanches, these rocks are often found on distal ocean shelves.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for identification & classification exercises
  • Useful for advanced study about sedimentary petrology
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.7 kg