Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531767 Supplier Invitrogen™ Supplier No. PA531767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (91%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRHL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene GRHL1
Gene Accession No. Q9NZI5
Gene Alias DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32
Gene Symbols GRHL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human GRHL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29841
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.