Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598889 Supplier Invitrogen™ Supplier No. PA598889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRHL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRHL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GRHL1
Gene Accession No. Q9NZI5
Gene Alias DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32
Gene Symbols GRHL1
Host Species Rabbit
Immunogen Recombinant Human Grainyhead-like protein 1 homologprotein (1-95AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29841
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.