Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRHL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$646.00
Specifications
Antigen | GRHL2 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence -Reported in scientific literature (PMID:24375798), Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GRHL2 Polyclonal specifically detects GRHL2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GRHL2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
79977 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence -Reported in scientific literature (PMID:24375798), Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
BOMMGC149294, Brother of mammalian grainyhead, deafness, autosomal dominant 28, DFNA28, FLJ13782, grainyhead-like 2 (Drosophila), grainyhead-like protein 2 homolog, TFCP2L3MGC149295, Transcription factor CP2-like 3FLJ11172 | |
GRHL2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title