Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRHL2 Monoclonal Antibody (CL3760)
GREENER_CHOICE

Catalog No. PIMA531388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531388 Supplier Invitrogen™ Supplier No. MA531388
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: NRVQVLKTVP VNLSLNQDHL ENSKREQYSI SFPESSAIIP VSGITVVKAE DFTPVFMAPP VHYPRGDGEE QRVVIFEQTQ YDVPSLATHS AYLKDDQRST PDSTYSESFK DAATEKFRSA SVGAEEYMYD QTSSGTFQYT LEATKSLRQK Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 87%.

The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRHL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL3760
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRHL2
Gene Accession No. Q6ISB3
Gene Alias 0610015A08Rik; BOM; Brother of mammalian grainyhead; DFNA28; ECTDS; grainyhead like 2; grainyhead like transcription factor 2; grainyheadlike; grainyhead-like 2; grainyhead-like 2 (Drosophila); grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; Grhl2; RGD1561191; Tcfcp2l3; TFCP2L3; transcription factor CP2-like 3
Gene Symbols GRHL2
Host Species Mouse
Immunogen Recombinant protein fragment of Human GRHL2 (Product #RP-102291)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.