Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587681 Supplier Invitrogen™ Supplier No. PA587681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSVSKASDSQ EDQEKRNCLG TSEAQSNLSG GENRVQVLKT VPVNLSLNQD HLENSKREQY SISFPESSAI IPVSGITVVK AEDFTPVFMA PPVHYPRGDG EEQRVVIFEQ TQYDVPSLAT HSAYLKDDQR STPDSTYSES FKDAATEKFR SASVGAEEYM YDQTSSGTFQ Y; Positive Samples: SH-SY5Y; Cellular Location: Membrane, Nucleus.

The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRHL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GRHL2
Gene Accession No. Q6ISB3
Gene Alias 0610015A08Rik; BOM; Brother of mammalian grainyhead; DFNA28; ECTDS; grainyhead like 2; grainyhead like transcription factor 2; grainyheadlike; grainyhead-like 2; grainyhead-like 2 (Drosophila); grainyhead-like protein 2 homolog; grainyhead-like transcription factor 2; Grhl2; RGD1561191; Tcfcp2l3; TFCP2L3; transcription factor CP2-like 3
Gene Symbols GRHL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-250 of human GRHL2 (NP_0791912).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299979, 79977
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.