Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRHPR Antibody (OTI2H4), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271430PECY55
Description
GRHPR Monoclonal specifically detects GRHPR in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| GRHPR | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| GRHPR | |
| Full length recombinant protein of human GRHPR (NP_036335) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI2H4 | |
| Flow Cytometry | |
| EC 1.1.1.81, GLXREC 1.1.1.79, glycerate-2-dehydrogenase, GLYD, glyoxylate reductase/hydroxypyruvate reductase, PH2 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 9380.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction