Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRHPR Monoclonal Antibody (OTI2H4)
GREENER_CHOICE

Catalog No. PIMA525338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525338 Supplier Invitrogen™ Supplier No. MA525338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRHPR Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRHPR
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2H4
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Grhpr
Gene Accession No. Q9UBQ7
Gene Alias 1110059D05Rik; 6430629L09Rik; Glxr; glycerate-2-dehydrogenase; GLYD; glyoxylate and hydroxypyruvate reductase; glyoxylate reductase/hydroxypyruvate reductase; GRHPR; MSTP035; PH2
Gene Symbols Grhpr
Host Species Mouse
Immunogen Full length human recombinant protein of GRHPR produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9380
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.