Learn More
GRHPR Mouse anti-Human, Clone: OTI10A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502090

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.Specifications
GRHPR | |
Monoclonal | |
Unconjugated | |
GRHPR | |
GLXR, GLYD, PH2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI10A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UBQ7 | |
GRHPR | |
Full length human recombit protein of human GRHPR produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.