Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRHPR Mouse anti-Human, Clone: OTI1H8, lyophilized, TrueMAB™
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF502095

Encompass_Preferred

Catalog No. 50-166-1032


May include imposed supplier surcharges.
Only null left
Add to cart

Description

Description

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.
Specifications

Specifications

GRHPR
Monoclonal
Unconjugated
GRHPR
GLXR, GLYD, PH2
Mouse
Affinity Chromatography
RUO
9380
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1H8
PBS with 8% trehalose and no preservative; pH 7.3
Q9UBQ7
GRHPR
Full length human recombit protein of human GRHPR produced in HEK293T cell.
100 μg
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.