Learn More
Invitrogen™ GRHPR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109787
Description
Immunogen sequence: VGYTPDVLTD TTAELAVSLL LTTCRRLPEA IEEVKNGGWT SWKPLWLCGY GLTQSTVGII GLGRIGQAIA RRLKPFGVQR FLYTGRQPRP EEAAEFQAEF VSTPELAAQS DFIVVACSLT PATEGLCNKD FFQKMKETAV FINISRGDVV NQDDLYQALA SGKIAAAGLD VTSPEPLPTN HPLLTLKNCV ILPHIGSATH RTRNTMSLLA ANNLLAGLRG EPMPSELKL.
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.
Specifications
| GRHPR | |
| Polyclonal | |
| Unconjugated | |
| Grhpr | |
| 1110059D05Rik; 6430629L09Rik; Glxr; glycerate-2-dehydrogenase; GLYD; glyoxylate and hydroxypyruvate reductase; glyoxylate reductase/hydroxypyruvate reductase; GRHPR; MSTP035; PH2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 680021, 76238, 9380 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.29 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91Z53, Q9UBQ7 | |
| Grhpr | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-328 of human GRHPR (NP_036335.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.