Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GRI 977143
GREENER_CHOICE

Catalog No. 477910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-791-0 10 mg
47-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-791-0 Supplier Tocris Bioscience™ Supplier No. 4779/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective LPA2 receptor non-lipid agonist

Selective lysophosphatidic acid 2 (LPA2) receptor non-lipid agonist (EC50 = 3.3 μM). Does not activate other LPA GPCRs at concentrations up to 10 μM. Antiapoptotic; inhibits activation of caspases 3, 7, 8 and 9, Bax translocation and PARP-1 cleavage. Activates ERK1/2 survival pathway.

Chemical Identifiers

CAS 325850-81-5
Molecular Formula C22H17NO4S

Specifications

Quantity 10 mg
Formula Weight 391.44
Purity >98%
Molecular Weight (g/mol) 391.441
Chemical Name or Material GRI 977143
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.