Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GRI 977143
GREENER_CHOICESDP

Catalog No. 477950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-795-0 50 mg
47-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-795-0 Supplier R&D Systems™ Supplier No. 4779/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective LPA2 receptor non-lipid agonist

Selective lysophosphatidic acid 2 (LPA2) receptor non-lipid agonist (EC50 = 3.3 μM). Does not activate other LPA GPCRs at concentrations up to 10 μM. Antiapoptotic; inhibits activation of caspases 3, 7, 8 and 9, Bax translocation and PARP-1 cleavage. Activates ERK1/2 survival pathway.

Chemical Identifiers

CAS 325850-81-5
Molecular Formula C22H17NO4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 391.44
Purity >98%
Molecular Weight (g/mol) 391.441
Chemical Name or Material GRI 977143
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.