Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRID2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298938C
Description
GRID2 Polyclonal specifically detects GRID2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GRID2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
B230104L07Rik, cpr, creeper, gluR delta-2 subunit, GluRdelta2, glutamate receptor delta-2 subunit, glutamate receptor, ionotropic, delta 2, ho, hotfoot, Lc, lurcher, minisatellite 10ac detected by probe MMS10, MMS10-AC, Ms10ac, nmf408, tpr | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GRID2. | |
0.1 mL | |
Neuronal Cell Markers, Neurotransmission | |
2895 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction