Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRID2IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560985 Supplier Invitrogen™ Supplier No. PA560985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SETSHMSVKR LRWEQVENSE GTIWGQLGED SDYDKLSDMV KYLDLELHFG TQKPAKPVPG PEPFRKKEVV EILSHKKAYN TSILLAHLKL SPAELRQVLM SMEP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRID2IP
Applications Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRID2IP
Gene Accession No. A4D2P6
Gene Alias Delphilin; GluR-delta2 philic-protein; glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein; glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein 1; glutamate receptor, ionotropic, delta 2-interacting protein 1; Grid2 interacting protein; GRID2IP; L-delphilin
Gene Symbols GRID2IP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRID2IP. Recombinant protein control fragment (Product #RP-100009).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.