Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Griffithsin Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144516 Supplier Invitrogen™ Supplier No. PA5144516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Griffithsin Polyclonal Antibody for ELISA, Western Blot

Mixed specificity lectin with anti-HIV activity. Binds to HIV envelope glycoproteins, including exterior membrane glycoprotein gp120, and inhibits viral entry into cells. Binding to gp120 is dependent on gp120 being glycosylated, and is inhibited by mannose, glucose and N-acetylglucosamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Griffithsin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Griffithsin
Host Species Rabbit
Immunogen Recombinant Griffithsia sp. Griffithsin protein (1-121aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Algae
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.