Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein
SDP

Catalog No. NB000014 Shop All R&D Systems Products
Change view
Click to view available options
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB000014 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000014 Supplier Novus Biologicals™ Supplier No. DL1208NB
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • A biologically active protein corresponding to Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4
  • Source: Griffonia (Bandeiraea) simplicifolia seeds
  • GSL I-B4 isolectin contains only the B subunits.
  • It is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit, and goat as well as a marker for non-peptidergic unmyelinated primary afferent neurons.
  • This “B”-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues.
  • DyLight™ 649 GSL I-B4 isolectin is produced by using special conjugation procedures to incorporate DyLight™ 649 into our affinity-purified lectin.
  • This conjugate has an appropriate number of fluorochromes bound which provide the maximum fluorescence and optimum staining characteristics for this particular lectin.
  • This reagent is supplied essentially free of unconjugated fluorochromes and inactive lectin.
  • Excitation maximum is at 646 nm and emission maximum is at 674 nm (red).

Specifications

For Use With (Application) Bioactivity, Immunocytochemistry/Immunofluorescence
Molecular Weight (g/mol) 114 kDa
Purification Method Affinity purified
Quantity 0.5 mg
Storage Requirements Store at 4°C in the dark.
Conjugate DyLight 649
Buffer 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca2+
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.