Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grifolic acid, R&D Systems™
SDP

Catalog No. 45811 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-811 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-811 Supplier R&D Systems Supplier No. 4581/1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective partial FFA4 (GPR120) agonist

Selective partial FFA4 (GPR120) agonist. Induces ERK and [Ca2+]i responses in cells expressing GPR120, but not those expressing GPR40.

Chemical Identifiers

CAS 80557-12-6
Molecular Formula C23H32O4
Molecular Weight (g/mol) 372.505
InChI Key QPIZDZGIXDKCRC-JTCWOHKRSA-N
Synonym grifolic acid, ilicicolinic acid b, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyldodeca-2,6,10-trienyl benzoic acid, d0e9ay, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecat rien-1-yl benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecatrien-1-yl-benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl benzoic acid, benzoic acid, 2,4-dihydroxy-6-methyl-3-2e,6e-3,7,11-trimethyl-2,6,10-dodecatrienyl
PubChem CID 9976563
IUPAC Name 2,4-dihydroxy-6-methyl-3-[(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl]benzoic acid
SMILES CC1=CC(=C(C(=C1C(=O)O)O)CC=C(C)CCC=C(C)CCC=C(C)C)O

Specifications

Quantity 1 mg
Formula Weight 372.5
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Grifolic acid
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.