Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRIK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145060
Description
Immunogen sequence: TEKAAGEVSK HLYKVWKKIG IWNSNSGLNM TDSNKDKSSN ITDSLANRTL Antigen sequence identity: Mouse 90%; Rat 90%.
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik1, also known as glutamate receptor 5, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Grik1 is expressed in GABAergic interneurons of the hippocampus and are thought to participate in the formation of various subtypes of kainate receptors with Grik2 and KA2. Stimulation of Grik1 leads to intracellular calcium release and activation of protein kinase C. Excessive activation has been associated with psychiatric, neurological and neurodegenerative diseases. Numerous isoforms of Grik1 are known to exist and may be subject to RNA editing within the second transmembrane domain, which is thought to alter the properties of ion flow.
Specifications
GRIK1 | |
Polyclonal | |
Unconjugated | |
Grik1 | |
A830007B11Rik; D16Ium24; D16Ium24e; EAA3; EEA3; excitatory amino acid receptor 3; GLR5; GluK1; GluK5; GLUR5; gluR-5; Glurbeta1; glutamate ionotropic receptor kainate type subunit 1; glutamate receptor; glutamate receptor 5; Glutamate receptor ionotropic, kainate 1; glutamate receptor subunit 5; glutamate receptor, ionotropic, kainate 1; GRIK1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P39086 | |
Grik1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 400-449. Recombinant protein control fragment (Product #RP-109925). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction