Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRIK4 Monoclonal Antibody (8H5G5)
GREENER_CHOICE

Catalog No. PIMA531745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531745 Supplier Invitrogen™ Supplier No. MA531745
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31745 has been tested in indirect ELISA.

This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIK4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8H5G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRIK4
Gene Accession No. Q16099
Gene Alias 6330551K01Rik; EAA1; Excitatory amino acid receptor 1; GluK4; GluRgamma1; glutamate ionotropic receptor kainate type subunit 4; glutamate receptor; glutamate receptor ionotropic, kainate 4; glutamate receptor KA1; Glutamate receptor KA-1; glutamate receptor, ionotropic, kainate 4; GRIK; Grik4; KA1; KA-1; kainate receceptor
Gene Symbols GRIK4
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIK4 (AA: extra 21-166) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.