Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIK4 Mouse anti-Human, Clone:8H5G5, Abnova™

Catalog No. 89333974 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-974 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-974 Supplier Abnova Corporation Supplier No. MAB17550
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human GRIK4.

This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members. [provided by RefSeq]

Specifications

Antigen GRIK4
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8H5G5
Conjugate Unconjugated
Description glutamate receptor, ionotropic, kainate 4
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene GRIK4
Gene Alias EAA1, GRIK, KA1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 21-166 of human GRIK4 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2900
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.