Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRIK5 Monoclonal Antibody (4G5H8)
GREENER_CHOICE

Catalog No. PIMA531747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531747 Supplier Invitrogen™ Supplier No. MA531747
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31747 has been tested in indirect ELISA.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik5, also known as kainate-preferring glutamate recptor subunit KA2, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Grik5 is highly homologous to the related ionotrophic glutamate receptor Grik4 (also known as KA1). Like Grik4, Grik5 does not form homomeric channels, but instead forms heteromers with Grik2. In Grik2- but not Grik1-null mice, Grik5 surface expression is greatly reduced in neurons, indicating that Grik2/Grik5 heteromers are required for exit from the endoplasmic reticulum to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIK5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G5H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRIK5
Gene Accession No. Q16478
Gene Alias EAA2; Excitatory amino acid receptor 2; GluK5; GluR gamma-2; GluRgamma2; glutamate ionotropic receptor kainate type subunit 5; glutamate receptor; glutamate receptor gamma-2; Glutamate receptor ionotropic, kainate 5; glutamate receptor KA2; glutamate receptor KA-2; glutamate receptor, ionotropic kainate 5; glutamate receptor, ionotropic, kainate 5; glutamate receptor, ionotropic, kainate 5 (gamma 2); GRIK2; GRIK5; iGlu5; KA2; LOW QUALITY PROTEIN: glutamate receptor ionotropic, kainate 5; MGC128958 protein
Gene Symbols GRIK5
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIK5 (AA: extra 21-166) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.