Learn More
Invitrogen™ Grim-19 Monoclonal Antibody (1A8), eBioscience™
Mouse Monoclonal Antibody
$314.65
Specifications
Antigen | Grim-19 |
---|---|
Clone | 1A8 |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Description
Description: This 1A8 monoclonal antibody reacts with human, mouse, and rat Grim-19. This 16 kDa protein is expressed in many human tissues, especially in the heart and skeletal muscle. Lower expression levels of Grim-19 can be observed in the liver, kidney, and brain. Originally described as a nuclear protein, reports indicate that Grim-19 is primarily localized to mitochondria as an essential component of complex I of the electron transport chain. Grim-19 expression can be induced by interferon-beta and all-trans-retinoic acid in tumor cell lines, leading to cell death. Finally, Grim-19 has been shown to bind the transactivation domain of STAT3 and inhibit its transcriptional activity. The molecular mechanisms of how Grim-19 regulates STAT3 within the cell remains under active investigation. Applications Reported: This 1A8 antibody has been reported for use in immunoblotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This 1A8 antibody has been tested by immunoblotting of HeLa cell lysate. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.Specifications
Grim-19 | |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9ERS2, Q9P0J0 | |
100911483, 314759, 51079, 67184 | |
Primary | |
Affinity chromatography |
1A8 | |
4°C | |
Monoclonal | |
Liquid | |
IgG2b | |
Human, Mouse, Rat | |
Ndufa13 | |
2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3 | |
Ndufa13, Ndufa13-ps1 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.