Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grin2a Mouse anti-Rat, HRP, Clone:S327A-38, Abnova™

Catalog No. 89334215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-215 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-215 Supplier Abnova Corporation Supplier No. MAB17347
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant rat Grin2a.

Specifications

Antigen Grin2a
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S327A-38
Conjugate HRP
Description glutamate receptor, ionotropic, N-methyl D-aspartate 2A
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Grin2a
Gene Alias NMDAR2A, NR2A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 75-325 at N-terminus of rat Grin2a.
Quantity 100 μg
Gene ID (Entrez) 24409
Target Species Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.