Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIN2B Mouse anti-Human, Mouse, Rat, Clone: 1C5E12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | GRIN2B |
---|---|
Clone | 1C5E12 |
Concentration | 0.66 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6984
![]() ![]() |
Proteintech Group Inc
665651IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer′s Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal domit 6-related mental retardation.Specifications
GRIN2B | |
0.66 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GluN2B, GRIN2B, hNR3, NMDAR2B, NR2B, NR3 | |
GRIN2B | |
IgG1 | |
Protein G | |
Antibody |
1C5E12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q00960, Q01097, Q13224 | |
14812, 24410, 2904 | |
GRIN2B Fusion Protein Ag16718 | |
Primary | |
-20°C | |
GRIN2B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title