Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRINA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534489 Supplier Thermo Scientific Supplier No. PA534489
Add to Cart
Add to Cart

GRINA Polyclonal Antibody

GRINA Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRINA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GRINA
Gene Accession No. Q7Z429
Gene Alias 1110025J15Rik; Glutamate [NMDA] receptor-associated protein 1; glutamate ionotropic receptor NMDA type subunit associated protein 1; glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding); glutamate receptor, NMDA subtype, glutamate-binding subunit; GRINA; HNRGW; Lag; LFG1; NMDA receptor glutamate-binding chain; NMDA receptor glutamate-binding subunit; Nmdara1; N-methyl-D-aspartate receptor glutamate-binding chain; Protein lifeguard 1; putative MAPK-activating protein PM02; TMBIM3; transmembrane BAX inhibitor motif containing 3; transmembrane BAX inhibitor motif-containing protein 3
Gene Symbols GRINA
Host Species Rabbit
Immunogen An 18 amino acid peptide near the center of human GRINA.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2907
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.