Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRINA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557896 Supplier Invitrogen™ Supplier No. PA557896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYAQPPYPGA PYPQPPFQPS PYGQPGYPHG PSPYPQGGYP QGPY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

The transmembrane BAX inhibitor motif (TMBIM) family of proteins includes the founder member TMBIM6/BI-1, TMBIM1/RECS1 (responsive to centrifugal force and shear stress gene 1 protein), TMBIM2/LFG (life guard), TMBIM3/GRINA (glutamate receptor ionotropic NMDA protein 1), TMBIM4/GAAP (Golgi anti-apoptotic-associated protein), and TMBIM5/GHTIM (growth hormone-inducible transmembrane protein). They are highly conserved in mammals and zebrafish and contain a conserved BAX inhibitor-1 motif. GRINA is expressed in the brain and is a potential apoptotic regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRINA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRINA
Gene Accession No. Q7Z429
Gene Alias 1110025J15Rik; Glutamate [NMDA] receptor-associated protein 1; glutamate ionotropic receptor NMDA type subunit associated protein 1; glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding); glutamate receptor, NMDA subtype, glutamate-binding subunit; GRINA; HNRGW; Lag; LFG1; NMDA receptor glutamate-binding chain; NMDA receptor glutamate-binding subunit; Nmdara1; N-methyl-D-aspartate receptor glutamate-binding chain; Protein lifeguard 1; putative MAPK-activating protein PM02; TMBIM3; transmembrane BAX inhibitor motif containing 3; transmembrane BAX inhibitor motif-containing protein 3
Gene Symbols GRINA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRINA. Recombinant protein control fragment (Product #RP-96582).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2907
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.