Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRINA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557897
Description
Immunogen sequence: PYGQPQVFPG QDPDSPQHGN YQEEGPPSYY DNQDFPATNW DDKSIRQAFI RK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
The transmembrane BAX inhibitor motif (TMBIM) family of proteins includes the founder member TMBIM6/BI-1, TMBIM1/RECS1 (responsive to centrifugal force and shear stress gene 1 protein), TMBIM2/LFG (life guard), TMBIM3/GRINA (glutamate receptor ionotropic NMDA protein 1), TMBIM4/GAAP (Golgi anti-apoptotic-associated protein), and TMBIM5/GHTIM (growth hormone-inducible transmembrane protein). They are highly conserved in mammals and zebrafish and contain a conserved BAX inhibitor-1 motif. GRINA is expressed in the brain and is a potential apoptotic regulator.
Specifications
GRINA | |
Polyclonal | |
Unconjugated | |
GRINA | |
1110025J15Rik; Glutamate [NMDA] receptor-associated protein 1; glutamate ionotropic receptor NMDA type subunit associated protein 1; glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding); glutamate receptor, NMDA subtype, glutamate-binding subunit; GRINA; HNRGW; Lag; LFG1; NMDA receptor glutamate-binding chain; NMDA receptor glutamate-binding subunit; Nmdara1; N-methyl-D-aspartate receptor glutamate-binding chain; Protein lifeguard 1; putative MAPK-activating protein PM02; TMBIM3; transmembrane BAX inhibitor motif containing 3; transmembrane BAX inhibitor motif-containing protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2907 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z429 | |
GRINA | |
Recombinant protein corresponding to Human GRINA. Recombinant protein control fragment (Product #RP-96581). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction