Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRINL1A Monoclonal Antibody (2E4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA522667

Catalog No. PIMA522667


Only null left
Add to Cart

Description

Description

Peptide Sequence: GSPISSEERR RRDKQHLDDI TAARLLPLHH MPTQLLSIEE SLALQKQQKQ NYEEMQAKLA AQKLAERLNI KMRSYNPEGE SSGRYREVRD EDDDWSSDEF.

This gene (Gcom1) is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts. The GRINL1A gene uses a downstream promoter for transcription and also has multiple alternatively spliced variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GRINL1A
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P0CAP2, Q6EEV4
POLR2M
POLR2M (NP_056347.1, 269 a.a. ~ 368 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
2E4
Unconjugated
POLR2M
AA407243; AA408799; AI303746; AW553238; D9Wsu138e; DNA-directed RNA polymerase II subunit GRINL1A; DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5; DNA-directed RNA polymerase II subunit M; DNA-directed RNA polymerase II subunit M, isoforms 4/5; gcom; GCOM1; Gcom2; Gdown; Gdown1; Glurr; glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A; glutamate receptor-like protein 1A; Grinl1a; GRINL1A downstream protein 1; GRINL1A downstream protein Gdown4; GRINL1A upstream protein; Gup; mKIAA4185; Myocardial zonula adherens protein; MYOZAP; MYZAP; MYZAP-POLR2M; POLR2M; polymerase (RNA) II (DNA directed) polypeptide M; polymerase (RNA) II subunit M; protein GRINL1A; protein GRINL1A, isoforms 4/5; RNA polymerase II subunit M
Mouse
Affinity Chromatography
RUO
81488
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.