Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRINL1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528472 Supplier Invitrogen™ Supplier No. PA528472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene (Gcom1) is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts. The GRINL1A gene uses a downstream promoter for transcription and also has multiple alternatively spliced variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRINL1A
Applications Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene POLR2M
Gene Accession No. P0CAP2, Q6EEV4
Gene Alias AA407243; AA408799; AI303746; AW553238; D9Wsu138e; DNA-directed RNA polymerase II subunit GRINL1A; DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5; DNA-directed RNA polymerase II subunit M; DNA-directed RNA polymerase II subunit M, isoforms 4/5; gcom; GCOM1; Gcom2; Gdown; Gdown1; Glurr; glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A; glutamate receptor-like protein 1A; Grinl1a; GRINL1A downstream protein 1; GRINL1A downstream protein Gdown4; GRINL1A upstream protein; Gup; mKIAA4185; Myocardial zonula adherens protein; MYOZAP; MYZAP; MYZAP-POLR2M; POLR2M; polymerase (RNA) II (DNA directed) polypeptide M; polymerase (RNA) II subunit M; protein GRINL1A; protein GRINL1A, isoforms 4/5; RNA polymerase II subunit M
Gene Symbols POLR2M
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human GRINL1A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.