Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241211MFV500
Description
GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| GRIP1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human GRIP1. The immunogen is located within the last 50 amino acids of GRIP1. Amino Acid Squence: QPSHGGNLETREPTNTL | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
| 23426 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction