Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1202842MFV610
Description
GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Hamster samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
GRIP1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1. | |
0.1 mL | |
Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
23426 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction