Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6653 Supplier Bethyl Laboratories, Inc Supplier No. A304004A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

Glutamate receptors play an important role in neural plasticity, development and degeneration. The glutamate receptor interacting proteins, GRIP1 and GRIP2, members of the PDZ domain-containing protein family, mediate the trafficking and membrane organization of a number of transmembrane proteins. GRIP1 and GRIP2 specifically bind to the AMPA receptor subunits, GluR 2/3 and are involved in the targeting of GluR 2/3 to the synapse. GRIP1 is expressed in early development before the expression of AMPA receptors, while GRIP2 expression parallels that of AMPA receptors during later developmental stages. GRIP1 and GRIP2 may be involved in the induction of cerebellar long-term depression (LTD).

Specifications

Antigen GRIP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene GRIP1
Gene Accession No. Q9Y3R0
Gene Alias GRIP, GRIP-1
Gene Symbols GRIP1
Host Species Rabbit
Immunogen Between 1026 and 1076
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23426
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.