Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733858 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3858 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3858 Supplier Proteintech Group Inc Supplier No. 223981AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRIP1 is involved in glutamate receptors, which play an important role in neural plasticity, development and degeneration. The glutamate receptor interacting proteins, GRIP1 and GRIP2, members of the PDZ domain-containing protein family, mediate the trafficking and membrane organization of a number of transmembrane proteins. GRIP1 and GRIP2 specifically bind to the AMPA receptor subunits, GluR 2/3 and are involved in the targeting of GluR 2/3 to the synapse. GRIP1 is expressed in early development before the expression of AMPA receptors, while GRIP2 expression parallels that of AMPA receptors during later developmental stages. GRIP1 and GRIP2 may be involved in the induction of cerebellar long-term depression (LTD).

Specifications

Antigen GRIP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GRIP1
Gene Accession No. Q925T6, Q9Y3R0
Gene Alias GRIP, GRIP1
Gene Symbols Grip1
Host Species Rabbit
Immunogen GRIP1 Fusion Protein Ag18071
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23426, 74053
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.