Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+)-Griseofulvin, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.
Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.
Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 218°C to 220°C |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 95226 |
Merck Index | 14,4549 |
Solubility Information | Soluble in dimethyl formamide,ethanol,methanol,chloroform,DMSO,tetrachloroethane. Insoluble in water. |
Formula Weight | 352.77 |
Percent Purity | 97% |
Chemical Name or Material | (+)-Griseofulvin |
Safety and Handling
Hazard Category | H317-H351-H360FD |
Hazard Statement | GHS H Statement H360-H351-H317 May damage fertility or the unborn child. Suspected of causing cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
EINECSNumber | 204-767-4 |
RTECSNumber | WG9800000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction