Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Griseofulvin, 97%, Thermo Scientific Chemicals

Catalog No. AC227451000
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-07-8
Molecular Formula C17H17ClO6
Molecular Weight (g/mol) 352.76
MDL Number MFCD00082343
InChI Key DDUHZTYCFQRHIY-RBHXEPJQSA-N
Synonym griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine
PubChem CID 441140
ChEBI CHEBI:27779
IUPAC Name (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
SMILES CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 217.0°C to 224.0°C
Color White to White-Yellow
Loss on Drying 1% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 4584
Specific Rotation Condition +354° (20°C c=1,dimethylform.)
Specific Rotation + 354.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dmf,dmso,slightly soluble in alcohol,benzene,acetone,,chloroform,ethyl acetate,practically insoluble in petroleum ether
Spectrophotometry 96.0% min. (on dried substance)
Formula Weight 352.76
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (+)-Griseofulvin, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
May cause an allergic skin reaction.
May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-767-4
RTECSNumber WG9800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.