Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK1 Antibody (D11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202776H
Description
GRK1 Monoclonal specifically detects GRK1 in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
GRK1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GRK1 | |
Full length human GRK1. | |
0.1 mL | |
Protein Kinase, Signal Transduction, Vision | |
6011 | |
Human, Bovine | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
D11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Detects G-protein-associated rhodopsin kinase 1a/b (GRK1a/b) from human and bovine tissues. This does not cross-react with other G protein-coupled kinases. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction