Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK1 Antibody (G8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202775AF532
Description
GRK1 Monoclonal specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GRK1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
GRK1 | |
Full length human GRK1. | |
Primary | |
Detects G-protein-associated rhodopsin kinase 1a (GRK1a). | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
G8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
Mouse | |
0.1 mL | |
6011 | |
Human, Mouse, Rat, Bovine, Chicken | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction