Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK1 Antibody (G8), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202775G
Description
GRK1 Monoclonal specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GRK1 | |
Monoclonal | |
DyLight 488 | |
G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Detects G-protein-associated rhodopsin kinase 1a (GRK1a). | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
G8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
GRK1 | |
Full length human GRK1. | |
0.1 mL | |
Protein Kinase, Signal Transduction, Vision | |
6011 | |
Human, Mouse, Rat, Bovine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction