Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRK1 Monoclonal Antibody (G8)
GREENER_CHOICE

Catalog No. MA1720
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1720 Supplier Invitrogen™ Supplier No. MA1720
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-720 detects G-protein-associated rhodopsin kinase 1a (GRK1a) from human, mouse, rat, bovine and chicken tissues. MA1-720 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ~60 kDa protein representing GRK1a from bovine rod outer segments. Immunofluorescence staining of GRK1a in bovine retina with MA1-720 results in intense staining primarily in cone outer segments, but also in rod outer segments. Weak staining is observed in somata and synaptic terminals of cones and inner segments of rods. The MA1-720 immunogen is full length human GRK1. This antibody recognizes an epitope in the C-terminal region of GRK1a.

GRK1 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinases. GRK1 mediates rapid desensitization of rod photoreceptors to light by catalyzing phosphorylation of the visual pigment rhodopsin which leads to its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). GRK1 can be phosphorylated by PKA and this reduces the ability of GRK1 to phosphorylate rhodopsin in vitro. It is speculated that phosphorylation of GRK1 by PKA in vivo occurs in the dark when cAMP levels in photoreceptor cells are elevated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRK1
Gene Accession No. P28327, Q15835, Q63651, Q9WVL4
Gene Alias G protein-coupled receptor kinase 1; G protein-coupled receptpr kinase 1; GPRK1; G-protein coupled receptor kinase 1; G-protein receptor kinase 1; GRK 1a/b; GRK1; GRK1a; rhodopsin kinase; rhodopsin kinase GRK1; Rhok; RK
Gene Symbols GRK1
Host Species Mouse
Immunogen Full length human GRK1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24013, 281457, 395431, 6011, 81760
Target Species Human, Mouse, Rat, Bovine, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.