Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRK2 Monoclonal Antibody (5D5)
GREENER_CHOICE

Catalog No. MA12013
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1-2013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1-2013 Supplier Invitrogen™ Supplier No. MA12013
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-2013 detects the beta andrenergic receptor kinase 1 in human, mouse and rat samples. MA1-2013 has been successfully used in Western blot, immunocytochemistry, immunofluoresence, immunoprecipitation, and ELISA procedures. By Western blot this antibody detects a ~30 kDa protein representing the beta andrenergeic receptor kinase 1 in human HeLa cells. In immunocytochemistry procedures MA1-2013 recognizes the beta andrenergic receptor kinase 1 in HeLa, Hek-293 and 3T3 cells. Immunofluoresence in HeLa cells yeilds predominantely cytoplasmic membrane associated staining with some staining in the ruffles. The MA1-2013 immunogen is recombinant beta adrenergic receptor kinase 1.

ADRBK1 (GRK2) is a serine/threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs). G protein-coupled receptor kinases (GRKs) are important regulators of G protein-coupled receptors(GPCRs). GRK2 (83 kDa), one of six members of this family that have been identified, is ubiquitously expressed in mammals. After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs (e.g., a2-adrenergic receptor) are phosphorylated by GRKs. Internalization of the GPCRs, regulated by beta-arrestin-1, leads to activation of the Ras - Raf - ERK1&2 signaling pathway. GRK2 activity is tightly controlled by different mechanisms including phosphorylation by kinases such as PKC, Src and ERK1&2, as well as interaction with various proteins. ERK phosphorylates and thus inactivates GRK2 on serine 670 in a negative feedback mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK2
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene GRK2
Gene Accession No. P25098, P26817, Q99MK8
Gene Alias ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; EC 2.7.1.126; G protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2; Grk2; GRK-2
Gene Symbols GRK2
Host Species Mouse
Immunogen Recombinant beta adrenergic receptor kinase 1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110355, 156, 25238
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.